• USA Home
  • P8391 - Phosphoglucose Isomerase from rabbit muscle

EMAIL THIS PAGE TO A FRIEND
P8391 Sigma

Phosphoglucose Isomerase from rabbit muscle

Type IV, ammonium sulfate suspension, 400-600 units/mg protein (E1%/280)

Synonym: D-Glucose-6-phosphate ketol-isomerase, PGI, Phosphosaccharomutase

Purchase

Description

Unit Definition

One unit will convert 1.0 μmole of D-fructose 6-phosphate to D-glucose 6-phosphate per min at pH 7.4 at 25 °C.

Physical form

Crystalline suspension in 2.3 M (NH4)2SO4 solution containing 25 mM imidazole, 1 mM EDTA and 0.1% 2-mercaptoethanol, pH 8.2

Price and Availability


All labs need water

Biomedical Applications
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?