• USA Home
  • P8406 - 6-Phosphogluconic Dehydrogenase from sheep liver

EMAIL THIS PAGE TO A FRIEND
P8406 Sigma

6-Phosphogluconic Dehydrogenase from sheep liver

lyophilized powder, 2-10 units/mg protein

Synonym: 6-Phospho-D-gluconate: NADP+ 2-oxidoreductase (decarboxylating)

Purchase

Properties

Related Categories 1.1.x.x Acting on hydroxyl groups, 1.x.x.x Oxidoreductases, Biochemicals and Reagents, Carbohydrate Metabolism, Cell Biology,
form   lyophilized powder
composition   Protein, ~80%
foreign activity   Hexokinase and glucose-6-phosphate dehydrogenase <0.3%
storage temp.   −20°C

Description

Analysis Note

Protein determined by biuret

Unit Definition

One unit will oxidize 1.0 μmole of 6-phospho-D-gluconate to D-ribulose 5-phosphate and CO2 per min at pH 7.4 at 37 °C in the presence of NADP+.

Physical form

Lyophilized powder containing Tris buffer

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

inhibitor

Product #

Description

Add to Cart

I1149 Iodoacetamide, BioUltra
I6125 Iodoacetamide, ≥99% (NMR), crystalline

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?