• USA Home
  • P8416 - Potassium phosphate monobasic

EMAIL THIS PAGE TO A FRIEND
P8416 Sigma

Potassium phosphate monobasic

plant cell culture tested, ≥99%

Synonym: Monopotassium phosphate, Potassium dihydrogen phosphate, prim.-Potassium phosphate

Purchase

Properties

Related Categories Biochemicals and Reagents, Biological Buffers, Buffers A to Z, Cell Culture, Chemical Synthesis,
InChI Key   GNSKLFRGEWLPPA-UHFFFAOYSA-M
assay   ≥99%
pKa (25 °C)   (1) 2.15, (2) 6.82, (3) 12.38 (phosphoric acid)
mp   252.6 °C(lit.)
density   2.338 g/mL at 25 °C(lit.)
suitability   plant cell culture tested

Description

Application

Use in plant culture media pH adjustment.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.




Filter out uncertainty.
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
TC6615500
Protocols & Articles

Related Content

BioUltra Biological Buffers

A buffer, as defined by Van Slyke [1], is "a substance which by its presence in solution increases the amount of acid or alkali that must be added to cause unit change in pH". Buffers are thus very i...
Keywords: Absorption, Adsorption, Biochemistry, Biological Buffers, Biological processes, Buffers, PAGE

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?