• USA Home
  • P8425 - Triethylene glycol monododecyl ether

EMAIL THIS PAGE TO A FRIEND
P8425 Sigma

Triethylene glycol monododecyl ether

≥98% (GC)

Synonym: C12E3, Dodecyl triethylene glycol ether, Dodecyltriglycol, Polyoxyethylene (3) lauryl ether

  • CAS Number 3055-94-5

  • Linear Formula CH3(CH2)11(OCH2CH2)3OH

  • Molecular Weight 318.49

  •  Beilstein Registry Number 1779029

  •  EC Number 221-280-2

  •  MDL number MFCD00042659

  •  PubChem Substance ID 24898946

Purchase

Properties

Related Categories Biochemicals and Reagents, Detergents, Detergents A to M, Non-Ionic Detergents
description   non-ionic
InChI Key   FKMHSNTVILORFA-UHFFFAOYSA-N
assay   ≥98% (GC)
density   0.927 g/mL at 20 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
KK7870000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?