• USA Home
  • P8443 - L-α-Phosphatidylinositol ammonium salt from bovine liver

EMAIL THIS PAGE TO A FRIEND
P8443 Sigma

L-α-Phosphatidylinositol ammonium salt from bovine liver

>98% (TLC), lyophilized powder

Synonym: 1,2-Diacyl-sn-glycero-3-phospho-(1-D-myo-inositol) ammonium potassium salt, PI, PtdIns

Purchase

Description

Biochem/physiol Actions

Phospholipid membrane component, precursor of inositol phosphates.

Other Notes

Contains primarily stearic and arachidonic acids.

Packaging

Packaged under Argon.

Price and Availability


Better Together

Discover your Target

The Future of Bioimaging
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?