EMAIL THIS PAGE TO A FRIEND
P8626 Sigma

PTH-alanine

Purchase

Properties

Related Categories Molecular Biology, Protein Sequencing, Protein Structural Analysis, Proteomics, Reagents for Protein Sequencing More...
InChI Key   XGPFPXALZVOKRI-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
RTECS 
MU2628000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?