• USA Home
  • P8632 - 5-Pregnene-3β,11β,17α,20β-tetrol

EMAIL THIS PAGE TO A FRIEND
P8632 Sigma

5-Pregnene-3β,11β,17α,20β-tetrol

Synonym: 3β,11β,17α,20β-Tetrahydroxy-5-pregnene

Purchase

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?