EMAIL THIS PAGE TO A FRIEND
P8657 Sigma

Phthalic acid

~99%

Synonym: 1,2-Benzenedicarboxylic acid

  • CAS Number 88-99-3

  • Linear Formula C6H4-1,2-(CO2H)2

  • Molecular Weight 166.13

  •  Beilstein Registry Number 608199

  •  EC Number 201-873-2

  •  MDL number MFCD00002467

  •  PubChem Substance ID 329820331

Purchase

Properties

InChI Key   XNGIFLGASWRNHJ-UHFFFAOYSA-N
assay   ~99%
mp   210-211 °C (dec.)(lit.)

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
TH9625000
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?