• USA Home
  • P8707 - L-α-Phosphatidyl-D-myo-inositol 4,5-diphosphate, dipalmitoyl

EMAIL THIS PAGE TO A FRIEND
P8707 Sigma

L-α-Phosphatidyl-D-myo-inositol 4,5-diphosphate, dipalmitoyl

  • Empirical Formula (Hill Notation) C41H81O19P3

  • Molecular Weight 970.99

  •  MDL number MFCD01861927

  •  PubChem Substance ID 329820333

Purchase

Properties

InChI Key   HKWJHKSHEWVOSS-MRQSADPDSA-N
solubility   DMSO: soluble
  chloroform: soluble
  ethanol: soluble
storage temp.   −20°C

Description

Substrates

Signaling lipid which apparently acts by binding to plekstrin homology domains; direct activator of inward rectifier potassium channels; phospholipase C and PI3-kinase substrate; activates phospholipase D.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?