• USA Home
  • P8742 - Posttranslational Modification of Proteins: Tools for Functional Proteomics
EMAIL THIS PAGE TO A FRIEND
P8742 Sigma
Posttranslational Modification of Proteins: Tools for Functional Proteomics
  •  ISBN-10 0-89603-678-2
  •  ISBN-13 978-0-89603-678-9
Purchase
Properties
Related Categories Books and Software, Labware, Proteomics Books More...
publication info   C. Kannicht, Humana Press, 2002, 385 pp., hard cover
mfr. no.   Humana
Description
General description
This volume describes reproducible methods for detecting and analyzing the posttranslational modifications of protein, particularly with regard to protein function, proteome research, and the characterization of pharmaceutical proteins. Methods include those for analyzing the assignment of disulfide bond sites in proteins, protein N-glycosylation and protein O-glycosylation, and oligosaccharides present at specific single glycosylation sites in a protein. Additional techniques facilitate the analysis of glycosylphosphatidylinositols, lipid modifications, protein phosphorylation and sulfation, protein methylation and acetylation, a-amidation and lysine hydroxylation
Table of Contents
Assignment of Disulfide Bonds in Proteins by Chemical Cleavage and Peptide Mapping by Mass Spectrometry
Carbohydrate Composition Analysis of Glycoproteins Using Highly Sensitive Fluorescence Detection Methods
Enzymatical Hydrolysis of N-Glycans from Glycoproteins and Fluorescent Labeling by 2-Aminobenzamide (2-AB)
Separation of N-Glycans by HPLC,
Enzymatic Sequence Analysis of N-Glycans
Immunological Detection of O-GlcNAc
Analysis of O-Glycosylation
Characterization of Site-Specific Glycosylation
Monitoring Glycosylation of Therapeutic Glycoproteins for Consistency Using Highly Fluorescent Anthranilic Acid
Metabolic Labeling and Structural Analysis of Glycosylphosphatidylinositols from Parasitic Protozoa
Analysis of S-Acylation of Proteins
Immunoblotting Methods for the Study of Protein Ubiquitination
Analysis of Methylation and Acetylation in E. coli Ribosomal Proteins
Identification of In Vivo Protein Phosphorylation Sites with Mass Spectrometry
Analysis of Tyrosine-O-Sulfation
a-Amidated Peptides: Approaches for Analysis
g-Glutamate and b-Hydroxyaspartate in Proteins
Detection of isoAspartate Residues as a Posttranslational Modification of Proteins and Peptides
Lysine Hydroxylation and Crosslinking of Collagen
Heterologous Expression in Endocrine Cells for Analysis of Posttranslational Modifications
2D-Electrophoresis: Detection of Glycosylation and Influence on Spot Pattern
Price and Availability
Detect Your Unseen Protein
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?