• USA Home
  • P8754 - 5β-Pregnane-3α,17α,21-triol-20-one

EMAIL THIS PAGE TO A FRIEND
P8754 Sigma

5β-Pregnane-3α,17α,21-triol-20-one

Synonym: 11-Deoxytetrahydrocortisol, 3α,17α,21-Trihydroxy-5β-pregnan-20-one, Tetrahydro-S, Tetrahydrocortexolone, Tetrahydrodeoxycortisol

Purchase

Description

Packaging

250 mg in poly bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?