• USA Home
  • P8757 - 4-Pregnene-17α,20β,21-triol-3,11-dione

EMAIL THIS PAGE TO A FRIEND
P8757 Sigma

4-Pregnene-17α,20β,21-triol-3,11-dione

Synonym: 17α,20β,21-Trihydroxy-4-pregnene-3,11-dione, 20β-Dihydrocortisone, Reichstein’s substance U

  • CAS Number 116-59-6

  • Empirical Formula (Hill Notation) C21H30O5

  • Molecular Weight 362.46

  •  PubChem Substance ID 329820336

Purchase

Properties

InChI Key   XBIDABJJGYNJTK-GNIMZFFESA-N

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?