• USA Home
  • P8812 - Phosphate-citrate buffer with urea hydrogen peroxide tablets-268 mg

EMAIL THIS PAGE TO A FRIEND
P8812 Sigma

Phosphate-citrate buffer with urea hydrogen peroxide tablets-268 mg

Purchase

Properties

storage temp.   2-8°C

Description

Other Notes

component of o-phenylenediamine tablet set - P-9187

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?