• USA Home
  • P8864 - Protein-Ligand Interactions: A Practical Approach
EMAIL THIS PAGE TO A FRIEND
P8864 Sigma
Protein-Ligand Interactions: A Practical Approach
Hydrodynamics and Calorimetry, Volume 1
  •  ISBN-10 0-19-963746-6
  •  ISBN-13 978-0-19-963746-1
Purchase
Properties
Related Categories Books and Software, Labware, Proteomics Books More...
publication info   S.E. Harding and B. Chowdhry, ed., Oxford University Press, 2001, 328 pp., soft cover
mfr. no.   Oxford University Press
Description
General description
This volume covers the principle hydrodynamic and calorimetric techniques for studying protein-ligand interactions. It includes equilibrium dialysis, rate dialysis, quantitative characterization by chromatography, sedimentation velocity and sedimentation equilibrium analytical ultracentrifugation for probing ligand binding, surface plasmon resonance, capillary electrophoresis, molecular electro-optics, high flux x-ray and neutron scattering, isothermal titration calorimetry and differential scanning microcalorimetry.
Table of Contents
List of protocols
1. Protein-ligand interactions and their analysis,
2. Equilibrium dialysis and rate dialysis,
3. Quantitative characterization of ligand binding by chromatography,
4. Sedimentation velocity analytical ultracentrifugation,
5. Sedimentation equilibrium in the analytical ultracentrifuge,
6. Surface plasmon resonance,
7. Capillary electrophoresis,
8. Molecular electro-optics,
9. High-flux x-ray and neutron solution scattering,
10. Isothermal titration calorimetry of biomolecules,
11. Differential scanning microcalorimetry,
List of suppliers
Index
Price and Availability
Detect Your Unseen Protein
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?