• USA Home
  • P8906 - Phosphorylase B, cross-linked from rabbit muscle

EMAIL THIS PAGE TO A FRIEND
P8906 Sigma

Phosphorylase B, cross-linked from rabbit muscle

for SDS Polyacrylamide Gel Electrophoresis

Synonym: Markers for high molecular weights: 100,000-500,000

Purchase

Properties

type   for SDS Polyacrylamide Gel Electrophoresis
mol wt   dimer mol wt 194,800 Da
  hexamer mol wt 584,400 Da
  monomer mol wt 97,400 Da
  pentamer mol wt 487,000 Da
  tetramer mol wt 389,600 Da
  trimer mol wt 292,000 Da
storage temp.   2-8°C

Description

Application

Not recommended for use in the Laemmli system.

Quality

Relative band intensity decreases as mol. wt. increases. Trace bands (heptamer through nonomer) may also be detected.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

S8320 SigmaMarker, high range, mol wt 36,000-200,000 Da

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?