• USA Home
  • P8972 - AutoPAGE Burst-Pouch™ 4.75% Acrylamide Solution for Automated Sequencing

EMAIL THIS PAGE TO A FRIEND
P8972 Sigma

AutoPAGE Burst-Pouch™ 4.75% Acrylamide Solution for Automated Sequencing

Purchase

Properties

storage temp.   2-8°C

Description

Quantity

Burst-Pouch prepares 60 ml of gel solution

Legal Information

AutoPAGE is a trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?