• USA Home
  • P9015 - 5α-Pregnane-3α,20α-diol

EMAIL THIS PAGE TO A FRIEND
P9015 Sigma

5α-Pregnane-3α,20α-diol

Synonym: 3α,20α-Dihydroxy-5α-pregnane, Allopregnane-3α,20α-diol

  • CAS Number 566-58-5

  • Empirical Formula (Hill Notation) C21H36O2

  • Molecular Weight 320.51

  •  MDL number MFCD00003679

  •  PubChem Substance ID 329820337

Purchase

Properties

InChI Key   YWYQTGBBEZQBGO-CGVINKDUSA-N

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?