• USA Home
  • P9138 - Pyrophosphatase, nucleotide from Crotalus atrox venom

EMAIL THIS PAGE TO A FRIEND
P9138 Sigma

Pyrophosphatase, nucleotide from Crotalus atrox venom

Type III, lyophilized powder, 4-8 units/mg protein

Synonym: Dinucleotide nucleotidohydrolase

Purchase

Properties

type   Type III
form   lyophilized powder
composition   Protein, ~80% biuret
packaging   vial of ~35 units
foreign activity   5′-nucleotidase <0.02 unit/mg protein
  Phosphodiesterase ~0.2 unit/mg protein
  alkaline phosphatase <0.002 unit/mg protein
storage temp.   −20°C

Description

Unit Definition

One unit will hydrolyze 1.0 μmole of β-NAD to NMN and AMP per min at pH 7.4 at 37°C in the presence of Mg ions.

Physical form

Lyophilized powder containing approx. 20% Tris buffer salts

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

P7383 Pyrophosphatase, Nucleotide from Crotalus adamanteus venom, Type II, lyophilized powder, 4-8 units/mg protein, vial of ~25 units

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?