EMAIL THIS PAGE TO A FRIEND
P9150 Sigma

Plasma

from goat

Synonym: Citrated plasma

Purchase

Properties

Related Categories Biochemicals and Reagents, Plasma, Plasma & Blood Proteins, Plasma, Blood, and Related Proteins and Reagents, Proteins and Derivatives More...
biological source   from goat
contains   3.8% trisodium citrate as anticoagulant
storage temp.   2-8°C

Description

Preparation Note

Whole blood collected with anticoagulants (9:1), pooled, and centrifuged. The resulting plasma is 0.45 μm filtered and lyophilized from the indicated volumes.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

P4639 Plasma, from bovine

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?