• USA Home
  • P9190 - Hanna pH Meter model HI 9124
EMAIL THIS PAGE TO A FRIEND
P9190 Sigma
Hanna pH Meter model HI 9124
0-14 pH, accuracy: 0.01, resolution: 0.01, 0-100 °C temperature, accuracy: 0.5 °C
Purchase
Properties
Related Categories Hand Held, Hanna, Hanna pH Meters, Hanna pH Meters and Accessories, Hanna pH model HI 9124,
mfr. no.   Hanna
measuring range   0-100 °C temperature, accuracy: 0.5 °C
  0-14 pH, accuracy: 0.01, resolution: 0.01
Description
General description
Microprocessor based pH/° C meter in waterproof casing.
Temperature: manual or automatic
• Probe included
• dual display
• Calibration: automatic one- or two-point
• display prompts for buffers
• preprogrammed with five NBS and European standard buffers
• READY indicator for accurate calibration
• Completely waterproof, totally immersible meter with rugged ABS case.
• Excellent for field
• last reading recall
• Automatic switch-off after 10 min (can be overridden)
• 20 min low battery indicator
• 500 hr use with 4 AAA batteries
Help in selecting the correct electrode can be viewed at Electrode Selection Guide
Hints on Electrode Maintenance
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?