• USA Home
  • P9194 - Finnpipette® Varichannel multichannel pipette
EMAIL THIS PAGE TO A FRIEND
P9194 Sigma
Finnpipette® Varichannel multichannel pipette
200-1000 μL, accuracy: 5.0 μL
Purchase
Properties
Related Categories Finn Pipettors, Labware, Liquid Handling Equipment, Pipettes, Pipettors and Tips, Pipettors More...
mfr. no.   Finn
measuring range   200-1000 μL, accuracy: 5.0 μL
Description
General description
The multichannel pipet with flexibility for many different pipetting applications. By installing individual channel assemblies, the pipet can be set up as anything from a one-channel to a twelve-channel pipet (maximum ten on the 200-1000 μl model), with individually variable spacing from 9 mm up (11mm on the 200-1000 μl model). All models are supplied with eight channel assemblies. Additional assemblies can be purchased separately.
Legal Information
Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?