• USA Home
  • P9222 - ProteinA–HyperD® F

EMAIL THIS PAGE TO A FRIEND
P9222 Sigma

ProteinA–HyperD® F

saline suspension with ethanol, 50 μm mean particle size

Synonym: Protein A–ceramic beads

Purchase

Properties

form   saline suspension with ethanol
mean particle size   50 μm
useful pH range   2 - 11
capacity   >30 mg/mL binding capacity (human or murine IgG)
storage temp.   2-8°C

Description

Features and Benefits

Characteristics are tolerance of very high flow rates without bed compression, and high exchange capacity. Stable to most common solvents.

Other Notes

HyperD media are highly porous, rigid ceramic beads filled with a derivatized hydrogel.

Physical form

Suspension in 1 M NaCl with 20% ethanol

Legal Information

HyperD is a registered trademark of Pall Corporation

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 1170 3/PG 3
WGK Germany 
3
Flash Point(F) 
105 °F
Flash Point(C) 
40 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?