• USA Home
  • P9285 - Poly(Lys, Trp) 4:1 hydrobromide

EMAIL THIS PAGE TO A FRIEND
P9285 Sigma

Poly(Lys, Trp) 4:1 hydrobromide

mol wt ≥20,000

Purchase

Properties

Related Categories Amino Acids, Biochemicals and Reagents, Polyamino Acids, Random Copolymers
mol wt   mol wt ≥20,000
storage temp.   −20°C

Description

Analysis Note

Molecular weight based on viscosity.

Other Notes

For additional technical information on polyamino acids please visit the Polyamino acid FAQ resource.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?