• USA Home
  • P9354 - Polypep® protein digest porcine

EMAIL THIS PAGE TO A FRIEND
P9354 Sigma

Polypep® protein digest porcine

for microbiology

Purchase

Properties

Related Categories Biochemicals and Reagents, Protein Extracts, Proteins and Derivatives More...
grade   for microbiology
viscosity   low(lit.)

Description

Legal Information

Polypep is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?