EMAIL THIS PAGE TO A FRIEND
P9376 Sigma

PTH-asparagine

Purchase

Properties

Related Categories Molecular Biology, Protein Sequencing, Protein Structural Analysis, Proteomics, Reagents for Protein Sequencing More...
InChI Key   ZQGVIKNSDQBZSS-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?