• USA Home
  • P9431 - Poly(Arg, Pro, Thr) hydrochloride

EMAIL THIS PAGE TO A FRIEND
P9431 Sigma

Poly(Arg, Pro, Thr) hydrochloride

Arg:Pro:Thr (6:3:1), mol wt 10,000-30,000

Purchase

Properties

Related Categories Amino Acids, Biochemicals and Reagents, Polyamino Acids, Random Copolymers
feed ratio   Arg:Pro:Thr (6:3:1)
mol wt   mol wt 10,000-30,000
impurities   <5% Ornithine residues
storage temp.   −20°C

Description

Analysis Note

Polymer composition may deviate from the molar feed ratios. Molecular weight based on viscosity of precursor ornithine copolymer.

Other Notes

For additional technical information on polyamino acids please visit the Polyamino acid FAQ resource.

Packaging

25 mg in poly bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Poly(Amino Acid) Block Copolymers for Drug Delivery and other Biomedical Applications

Carmen Scholz Department of Chemistry, University of Alabama in Huntsville Huntsville, AL 35899 Email: scholzc@uah.edu
Keywords: Addition reactions, Asymmetric synthesis, Building blocks, Catalysis, Degradations, Living polymerization, PEGylations, Polymerization reactions, Precipitation, Proteomics, Ring-opening polymerization, Solvents

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?