• USA Home
  • P9482 - Anti-Potassium Channel Kv1.6 (Voltage-gated, Delayed-Rectifier Potassium Channel; RCK2; KV2) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
P9482 Sigma

Anti-Potassium Channel Kv1.6 (Voltage-gated, Delayed-Rectifier Potassium Channel; RCK2; KV2) antibody produced in rabbit

IgG fraction of antiserum, lyophilized powder

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to Ion Channels, Antibodies to Potassium Channels,
species reactivity   rat, mouse
application(s)   immunocytochemistry: 1:50 using cells permeabilized with 0.2% Triton X-100
  western blot (chemiluminescent): 1:100
clone   polyclonal
antibody form   IgG fraction of antiserum
form   lyophilized powder
storage temp.   −20°C
Gene Information   mouse ... Kcna6(16494)
rat ... Kcna6(64358)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

highly purified GST fusion protein from the C-terminal of rat potassium channel Kv1.6. The protein corresponds to amino acids 463-530.

Specificity

Recognizes full-length Kv1.6 protein. Does not cross-react with other potassium channel antigens.

Preparation Note

Protein A purified.

Price and Availability


Antibody Explorer

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?