• USA Home
  • P9491 - Hanna hand-held thermometer
EMAIL THIS PAGE TO A FRIEND
P9491 Sigma
Hanna hand-held thermometer
HI 765 probe for meter only
Purchase
Properties
Related Categories Electronic Meters, Hanna, Hanna Hand-Held Thermometer, Hanna Temperature measurement, Labware,
material   (Probe: Stainless Steel AISI 316/3.9 in. × 0.12 in. (100 mm × 3 mm))
  blue (handle color)
mfr. no.   Hanna
parameter   -58-302 °F temperature
Description
General description
Cable length: 3.3 ft. (1 m); probe handle: ABS; Probes are supplied with a possible error of 0.8 ºF (0.4 ºC): T.C.: 21 seconds
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
related product
Product #
Description
Add to Cart
92451 Timestrip Plus 25 °C
08168 Timestrip Plus 8 °C
06693 Timestrip Plus -20 °C
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?