EMAIL THIS PAGE TO A FRIEND
P9501 Sigma

PTH-aspartic acid

Purchase

Properties

InChI Key   NFUCNAREVSTFNC-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
NI8720800

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?