• USA Home
  • P9532 - Bis(polyethylene glycol bis[imidazoyl carbonyl])

EMAIL THIS PAGE TO A FRIEND
P9532 Sigma

Bis(polyethylene glycol bis[imidazoyl carbonyl])

Synonym: Bis(polyoxyethylene bis[imidazoyl carbonyl])

Purchase

Properties

Related Categories Biochemicals and Reagents, Crosslinking, Detergents, Detergents A to M, Detergents by Applications,
description   non-ionic
form   powder
solubility   H2O: ≥10 mg/mL
  methylene chloride: ≤30 mg/mL
storage temp.   −20°C

Description

Application

A homobifunctional, long chain cross-linking reagent. Reacts with primary amines resulting in a moderately stable carbamate linkage. Variations of this compound have been used to prepare protein conjugates.

Preparation Note

Prepared from PEG Compound, a tetravalent polyethylene glycol derivative, M.W. approx. 20,000.

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?