• USA Home
  • P9616 - Hanna pH/ORP/ °C meter model HI 98150

EMAIL THIS PAGE TO A FRIEND
P9616 Sigma

Hanna pH/ORP/ °C meter model HI 98150

size 220 mm × 82 mm × 66 mm

Purchase

Properties

Related Categories Labware, Portable Meters, pH Meters, pH Supplies
mfr. no.   Hanna
size   220 mm × 82 mm × 66 mm
weight   500 g
battery power   4 × 1.5 V AA, 350 hr life (continuous use without backlight or 12 V DC adapter included)

Description

General description

Waterproof, portable, printing and logging pH/ORP/°C meter with GLP built-in printer supplied with HI 1618D intelligent electrode with built-in temp. sensor, DIN, 1 m (3.3 ft) cable, 12VDC adapter, pH 4.01/7.01 buffer solutions, cleaning sol., 5 rolls of printer paper, batteries, hard carrying case.
Range, resolution: -4-19.99 ± 0.01pH; ±400.0 ± 0.1 (ISE), ±2000 ± 1 (ORP)
Temperature compensation: automatic from -10-120 °C (14-248 °F)
Calibration: automatic 1 or 2 points with 5 memorized buffers (pH 4.01, 6.86, 7.01, 9.18, 10.01)
Features: uses plain paper; extended pH range; intelligent electrode technology; store up to 500 measurements; dual-level backlit LCD; GLP capabilities

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?