EMAIL THIS PAGE TO A FRIEND
P9634 Sigma

Palmitoleyl alcohol

~99%

Synonym: cis-9-Hexadecenol

Purchase

Properties

InChI Key   LBIYNOAMNIKVKF-FPLPWBNLSA-N
assay   ~99%
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
nwg

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?