EMAIL THIS PAGE TO A FRIEND
P9652 Sigma

Polybuffer® 74

Purchase

Description

Application

Polybuffer is used in chromatography reagents and solvents. Polybuffer has been used to develop protocols for typical chromatofocusing experiments as well as to purifiy and characterize β-mannosidases from white rot fungus Phlebia radiata. Polybuffer has also been used to isolate isoforms of apolipoprotein CIII from human serum by chromatofocusing.

Legal Information

Polybuffer is a registered trademark of GE Healthcare

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?