• USA Home
  • P9732 - Anti-Potassium Channel Kv3.1b antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
P9732 Sigma

Anti-Potassium Channel Kv3.1b antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to Ion Channels, Antibodies to Potassium Channels,
species reactivity   mouse, rat, monkey, canine
application(s)   immunohistochemistry (frozen sections): suitable
  immunoprecipitation: suitable
  western blot (chemiluminescent): 1:200
clone   polyclonal
antibody form   affinity isolated antibody
form   lyophilized powder
storage temp.   −20°C
Gene Information   rat ... Kcnc1(25327)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to amino acids 567-585 of rat potassium channel Kv3.1b. This epitope is not present in the alternatively spliced Kv3.1a form.

Physical form

Lyophilized at ~0.8 mg/ml from phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin, 5% sucrose and 0.025% sodium azide.

Specificity

Recognizes full-length Kv3.1b protein.

Price and Availability


Antibody Explorer

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?