• USA Home
  • P9772 - 5-Pregnene-3β,20α-diol diacetate

EMAIL THIS PAGE TO A FRIEND
P9772 Sigma

5-Pregnene-3β,20α-diol diacetate

Synonym: 3β,20α-Diacetoxy-5-pregnene, 3β,20α-Dihydroxy-5-pregnene 3,20-diacetate

Purchase

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?