• USA Home
  • P9799 - pGlu-Gly-Arg-Phe amide acetate salt

EMAIL THIS PAGE TO A FRIEND
P9799 Sigma

pGlu-Gly-Arg-Phe amide acetate salt

≥97% (HPLC)

Synonym: Antho-RFamide

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Other Peptides, Peptides and Proteins, Peptides for Cell Biology More...
InChI Key   CMWHGNOBFVLNTC-UHFFFAOYSA-N
assay   ≥97% (HPLC)
storage temp.   −20°C
Gene Information   mouse ... Nts(67405)

Description

Biochem/physiol Actions

May be a neurotransmitter at neuromuscular junctions.

Other Notes

Neuropeptide originally isolated from the pennatulid Renilla koellikeri and the sea anemone Anthopleura elegantissima

Price and Availability


Amplified Detection

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?