• USA Home
  • P9824 - Filter membranes, Isopore track-etched polycarbonate
EMAIL THIS PAGE TO A FRIEND
P9824 Sigma
Filter membranes, Isopore track-etched polycarbonate
pore size 5.0 μm, diam. 47 mm
Purchase
Properties
Related Categories Filter membranes, Isopore track-etched polycarbonate, Filtration Supplies, Labware, Manufacturer Browser, Membrane Filters,
material   white
mfr. no.   Millipore TMTP04700
  Millipore, TMTP04700
diam.   47 mm
thickness   7-22 μm
pore size   5.0 μm pore size
Description
General description
Unique white membrane for collection of particulates for inspection by light or electron microscopy. Isopore membranes have a flat, glassy, non-staining surface. They are hydrophilic, with a PVP wetting agent, and autoclavable.
Manufacturing process produces a membrane with precise pore size and narrow pore size distribution for accurate separations by particle size.
Price and Availability
Bioburden Testing
Sterifil Aseptic System and Holder
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?