• USA Home
  • P9849 - Pepsin, cGmp From Porcine Stomach Mucosa

EMAIL THIS PAGE TO A FRIEND
P9849 Sigma

Pepsin, cGmp From Porcine Stomach Mucosa

Purchase

Properties

storage temp.   −20°C

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
SC6132000

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?