• USA Home
  • P9898 - 1,2-Diarachidoyl-sn-glycero-3-phosphocholine

EMAIL THIS PAGE TO A FRIEND
P9898 Sigma

1,2-Diarachidoyl-sn-glycero-3-phosphocholine

≥99% (TLC)

Synonym: L-α-Phosphatidylcholine, diarachidoyl

  • CAS Number 61596-53-0

  • Empirical Formula (Hill Notation) C48H96NO8P

  • Molecular Weight 846.25

  •  MDL number MFCD00067533

  •  PubChem Substance ID 24898929

Purchase

Properties

Related Categories Biochemicals and Reagents, Cell Biology, Glycerophospholipid Metabolism, Glycerophospholipids, Glycerophosphonocholines,
InChI Key   YHKJGWQODTZLHJ-UHFFFAOYSA-N
assay   ≥99% (TLC)
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?