• USA Home
  • P9903 - Pancreatic Polypeptide human

EMAIL THIS PAGE TO A FRIEND
P9903 Sigma

Pancreatic Polypeptide human

≥95% (HPLC)

Synonym: Ala-Pro-Leu-Glu-Pro-Val-Tyr-Pro-Gly-Asp-Asn-Ala-Thr-Pro-Glu-Gln-Met-Ala-Gln-Tyr-Ala-Ala-Asp-Leu-Arg-Arg-Tyr-Ile-Asn-Met-Leu-Thr-Arg-Pro-Arg-Tyr-NH2

  • CAS Number 75976-10-2

  • Empirical Formula (Hill Notation) C185H287N53O54S2

  • Molecular Weight 4181.71

  •  MDL number MFCD00081812

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Gastrointestinal Peptides, Natriuretic Peptides, Pancreatic Polypeptides,
assay   ≥95% (HPLC)
storage temp.   −20°C
Gene Information   human ... PPY(5539)

Description

Amino Acid Sequence

Ala-Pro-Leu-Glu-Pro-Val-Tyr-Pro-Gly-Asp-Asn-Ala-Thr-Pro-Glu-Gln-Met-Ala-Gln-Tyr-Ala-Ala-Asp-Leu-Arg-Arg-Tyr-Ile-Asn-Met-Leu-Thr-Arg-Pro-Arg-Tyr-NH2

Biochem/physiol Actions

Agonist at Y4 neuropeptide Y receptors.

Price and Availability


Amplified Detection

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?