• USA Home
  • P9905 - Polyacrylamide hydrazide

EMAIL THIS PAGE TO A FRIEND
P9905 Sigma

Polyacrylamide hydrazide

suitable for immobilization of microbial cells by enmeshing

Purchase

Properties

extent of labeling   ~900 μmol per g
matrix   Linear polyacrylamide (non-cross-linked; water-soluble)
  linear polyacrylamide (non-cross-linked; water soluble)
suitability   suitable for immobilization of microbial cells by enmeshing
storage temp.   −20°C

Description

Application

Cells are suspended in a solution of linear, water-soluble polymer, which is then gelled by addition of a suitable cross-linking agent (e.g., glyoxal).

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?