• USA Home
  • P9957 - L-Phenylalanine decarboxylase from Streptococcus faecalis

EMAIL THIS PAGE TO A FRIEND
P9957 Sigma

L-Phenylalanine decarboxylase from Streptococcus faecalis

dried cells, ≥5 units/g solid

Synonym: L-Phenylalanine carboxy-lyase

Purchase

Description

Biochem/physiol Actions

Decarboxylates phenylalanine to phenethylamine and tyrosine to tyramine

Unit Definition

One unit will liberate 1.0 μmole of CO2 from L-phenylalanine per min at pH 5.5 at 37°C.

Physical form

Dried cells from which activity can be extracted

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?