• USA Home
  • P9983 - Phorbol 12,13-didecanoate 20-homovanillate

EMAIL THIS PAGE TO A FRIEND
P9983 Sigma

Phorbol 12,13-didecanoate 20-homovanillate

≥98%

Synonym: PDDHV

  • Empirical Formula (Hill Notation) C49H72O11

  • Molecular Weight 837.09

  •  MDL number MFCD02684413

  •  PubChem Substance ID 24899066

Purchase

Properties

Related Categories Bioactive Small Molecule Alphabetical Index, Bioactive Small Molecules, Cell Biology, Cell Signaling and Neuroscience, Neurobiology,
InChI Key   DXGOIXWTPMLJIK-FPBOOSLESA-N
assay   ≥98%
solubility   DMSO: soluble
  H2O: insoluble
  chloroform: soluble
  ethanol: soluble
  methanol: soluble
storage temp.   −20°C
Gene Information   rat ... Trpv1(83810)

Description

Biochem/physiol Actions

Resiniferatoxin-type vanilloid phorbol ester with capsaicin-like selectivity for the vanilloid receptor.

Legal Information

Sold in France, Germany, Great Britain and Switzerland under a patent license, restricted to research use only, from Procyon Pharmaceuticals, Inc.

Price and Availability


Better Together

Discover your Target

Making Headway
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?