• USA Home
  • PAD15 - N-Phenyl-p-phenylenediamine

EMAIL THIS PAGE TO A FRIEND
PAD15 Sigma

N-Phenyl-p-phenylenediamine

capsule (15 mg)

Synonym: 4-Aminodiphenylamine, Variamine Blue RT Base

Purchase

Properties

InChI Key   ATGUVEKSASEFFO-UHFFFAOYSA-N
form   capsule (15 mg)
mp   68-72 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3
RTECS 
ST3150000
Flash Point(F) 
379.4 °F
Flash Point(C) 
193 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

241393 N-Phenyl-p-phenylenediamine, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?