• USA Home
  • PBS1 - Phosphate Buffered Saline System
EMAIL THIS PAGE TO A FRIEND
PBS1 Sigma
Phosphate Buffered Saline System
Purchase
Description
Packaging
Set of stock solutions for preparing custom PBS buffers
Price and Availability
Interactive Buffer Table
Standard component
Description
Product #
Add to Cart
1M KH2PO4 1 L SDS    
1M K2HPO4 1 L SDS    
5M NaCl 1 L SDS    
Safety & Documentation
Safety Information
RIDADR 
NONH for all modes of transport
WGK Germany 
1
Documents
Certificate of Analysis
Protocols & Articles
Related Content
BioUltra Biological Buffers
A buffer, as defined by Van Slyke [1], is "a substance which by its presence in solution increases the amount of acid or alkali that must be added to cause unit change in pH". Buffers are thus very i...
Keywords: Absorption, Adsorption, Biochemistry, Biological Buffers, Biological processes, Buffers, PAGE
Peer-Reviewed Papers
15
References
Set your institution to view full text papers.
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?