• USA Home
  • PG1000200 - Controlled pore glass–uncoated
EMAIL THIS PAGE TO A FRIEND
PG1000200 Sigma
Controlled pore glass–uncoated
nominal diameter 1,000 Å, 120-200 mesh
Synonym: Glass, controlled-pore
Purchase
Properties
Related Categories Analytical/Chromatography, CPR - Controlled Pore Glass, Gel Filtration, LPLC Separation Media, LPLC/TLC/Paper Chromatography More...
particle size   120-200 mesh
pore size   1,000 Å nominal diameter
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?