• USA Home
  • PG120200 - Controlled pore glass–uncoated

EMAIL THIS PAGE TO A FRIEND
PG120200 Sigma

Controlled pore glass–uncoated

nominal diameter 120 Å, 120-200 mesh

Synonym: Glass, controlled-pore

Purchase

Properties

Related Categories Analytical/Chromatography, CPR - Controlled Pore Glass, Gel Filtration, LPLC Separation Media, LPLC/TLC/Paper Chromatography More...
particle size   120-200 mesh
pore size   120 Å nominal diameter

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?