• USA Home
  • PIKSI - PIKSI-M Mimetic screen kit

EMAIL THIS PAGE TO A FRIEND
PIKSI Sigma

PIKSI-M Mimetic screen kit

Purchase

Properties

Related Categories Affinity Chromatography, Dye Resins, Molecular Biology, Protein Chromatography, Proteomics More...
shipped in   wet ice
storage temp.   2-8°C

Description

Components

Contains 1mL columns of the following chromatographic media
Mimetic Red® 2 A6XL
Mimetic Red® 3 A6XL
Mimetic Orange® 1 A6XL
Mimetic Orange® 2 A6XL
Mimetic Orange® 3 A6XL
Mimetic Yellow® 1 A6XL
Mimetic Yellow® 2 A6XL
Mimetic Green® 1 A6XL
Mimetic Blue® 1 P6XL
Mimetic Blue® SA P6XL

Application

Mimetic columns are used in protein chromatography, affinity chromatography and dye resins. Mimetic columns have been used in studies to develop anti-inflammatory therapeutics.

Legal Information

Mimetic Blue is a registered trademark of PROMETIC BIOSCIENCES, LTD.

Mimetic Green is a registered trademark of PROMETIC BIOSCIENCES, LTD.

Mimetic Orange is a registered trademark of PROMETIC BIOSCIENCES, LTD.

Mimetic Red is a registered trademark of PROMETIC BIOSCIENCES, LTD.

Mimetic Yellow is a registered trademark of PROMETIC BIOSCIENCES, LTD.

Price and Availability


Detect Your Unseen Protein
Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
RIDADR 
UN 1993C 3 / PGIII
WGK Germany 
3
Flash Point(F) 
95 °F
Flash Point(C) 
35 °C

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?