• USA Home
  • PROT20S - ProteoPrep® 20 Plasma Immunodepletion Kit Single

EMAIL THIS PAGE TO A FRIEND
PROT20S Sigma

ProteoPrep® 20 Plasma Immunodepletion Kit Single

Purchase

Properties

Related Categories Molecular Biology, Native Protein Sample Preparation, Protein Depletion, Proteomics
storage temp.   2-8°C

Description

Application

The ProteoPrep 20 Plasma Immunodepletion Kit specifically removes 20 of the most abundant proteins from human plasma or serum in preparation for further proteomics analysis. This depletion enables deeper penetration into the plasma proteome whether you use one- or two-dimensional electrophoresis, single or multidimensional chromatography or go straight to mass spectrometry. This is based on a unique conjugation process using small recombinant immunoaffinity ligands and conventional antibodies that permits high density conjugation on the support. The ligand selection ensures optimal specificity. Typical depletions remove 97-98% of the total protein mass in human plasma or serum.

Features and Benefits

• Increase your ability to visualize low abundance proteins and biomarkers - Removal of >99% of twenty of the most abundant proteins allows up to 50-fold increased protein loads, enhancing opportunities to visualize low abundance proteins

• Unique antibody affinity media - Novel high density antibody media displays higher specificity, reducing contamination and significantly improving depletion capacity compared to any currently available products

• Convenient spin-column format - Fast depletion and recovery accelerates throughput and validation making this the preferred choice for all proteomics labs

Legal Information

ProteoPrep is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability


Ultrafree Centrifugal Filter Units

Kit component also available separately

Description

Product #

Add to Cart

Ultrafree-MC microcentrifuge filters, MWCO 5,000 Da, PLCC cellulosic (regenerated cellulose) membrane SDS M0286

Kit component only

Description

Product #

Add to Cart

ProteoPrep® 20 Immunodepletion Column SDS    
ProteoPrep® 20 Equilibration Buffer, 10× concentrate SDS    
ProteoPrep® 20 Elution Solution , 10× concentrate SDS    
ProteoPrep® Preservative Concentrate SDS    
Luer-Lock Caps SDS    
Collection Tubes SDS    
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?