EMAIL THIS PAGE TO A FRIEND
PZ0182 Sigma

Pactamycin

≥95% (HPLC)

Synonym: 2-Hydroxy-6-methylbenzoic acid [(1S,2R,3R,4S,5S)-5-[(3-acetylphenyl)amino]-4-amino-3-[[(dimethylamino)carbonyl]amino]-1,2-dihydroxy-3-[(1S)-1-hydroxyethyl]-2-methylcyclopentyl]methyl ester, PNU-0015800, U-15800

Purchase

Properties

Related Categories Approved Therapeutics/Drug Candidates, Bioactive Small Molecule Alphabetical Index, Bioactive Small Molecules, Cell Biology, Cell Signaling and Neuroscience,
InChI Key   WVIUOSJLUCTGFK-JUJPXXQGSA-N
assay   ≥95% (HPLC)
form   powder
optical activity   [α]/D +20 to +30° in 95% ethanol
color   white to tan
solubility   DMSO: ≥12 mg/mL
originator   Pfizer
storage temp.   2-8°C

Description

Other Notes

This compound was developed by Pfizer for Immunology research. To learn more about Sigma′s partnership with Pfizer and view other authentic, high-quality Pfizer compounds, visit sigma.com/bsm-pfizer.

To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Legal Information

Sold for research purposes under agreement from Pfizer Inc.

LOPAC is a registered trademark of Sigma-Aldrich Co. LLC

Biochem/physiol Actions

Pactamycin is a potent protein synthesis inhibitor, inhibiting protein synthesis at the translocation step on the 70S ribosome. It has activity against Gram-positive and Gram-negative bacteria, and broad antitumor, antiviral, and antiplasmodial activity. Cytotoxicity limits it clinical use.

Features and Benefits

This compound is also offered as part of Sigma′s Library of Pharmacologically Active Compounds (LOPAC®Pfizer), a collection of 90 Pfizer-developed drugs and research tools. Click here to learn more.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGII
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles

Articles

Discover Bioactive Small Molecules for Immunology Research

The immune system is a complex network to detect and defend against non-self pathogenic agents, such as disease-causing viruses, bacteria, and other microorganisms. Various signaling pathways contrib...
Keywords: Antibiotics, Antifungals, Antivirals, Apoptosis, Bioactive small molecules, Cell signaling, Diabetes, Diseases, Immunology, Inflammation

Pfizer Compounds for Immunology

From our library of Articles, Sigma-Aldrich presents Pfizer Compounds for Immunology

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?